accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNO

CAS RN 27258-33-9

IUPAC Name: 1-methyl-1H-pyrazole-5-carbaldehyde
Synonyms: 2-methylpyrazole-3-carbaldehyde
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: RAJRANFZSWDUJZ-UHFFFAOYSA-N
SMILES: CN1N=CC=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results