CAS RN 27300-28-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Acetyl-3,4,5,6-tetrahydropyridine Hydrochloride(Technical Grade), TRC
CAS: 27300-28-3 Molecular Formula: C7H11NO. HCl Molecular Weight (g/mol): 161.63 Synonym: 1-(3,4,5,6-Tetrahydro-2-pyridinyl)ethanone Hydrochloride,Methyl 3,4,5,6 Tetrahydro-2-pyridyl Ketone Hydrochloride,1-(2,3,4,5-tetrahydropyridin-6-yl)ethanone;hydrochloride, 2-Acetyl-3,4,5,6-tetrahydropyridine Hydrochloride IUPAC Name: 1-(2,3,4,5-tetrahydropyridin-6-yl)ethanone;hydrochloride SMILES: CC(=O)C1=NCCCC1.Cl