missing translation for 'onlineSavingsMsg'
Learn More

2-Acetyl-3,4,5,6-tetrahydropyridine Hydrochloride(Technical Grade), TRC

Product Code. 30134067
Change view
Click to view available options
Quantity:
10 mg
100 mg
250 mg
Unit Size:
100mg
10mg
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134067 250 mg 250mg
30133261 100 mg 100mg
17948144 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134067 Supplier Toronto Research Chemicals Supplier No. TRCA188500250MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27300-28-3
Molecular Formula C7H11NO. HCl
Molecular Weight (g/mol) 161.63
Synonym 1-(3,4,5,6-Tetrahydro-2-pyridinyl)ethanone Hydrochloride, Methyl 3,4,5,6 Tetrahydro-2-pyridyl Ketone Hydrochloride, 1-(2,3,4,5-tetrahydropyridin-6-yl)ethanone;hydrochloride, 2-Acetyl-3,4,5,6-tetrahydropyridine Hydrochloride
IUPAC Name 1-(2,3,4,5-tetrahydropyridin-6-yl)ethanone;hydrochloride
SMILES CC(=O)C1=NCCCC1.Cl

Specifications

Quantity 250 mg
InChI Formula InChi=1S/C7H11NO.ClH/c1-6(9)7-4-2-3-5-8-7;/h2-5H2,1H3;1H
Formula Weight 161.06
Chemical Name or Material 2-Acetyl-3,4,5,6-tetrahydropyridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.