accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2745-49-5

ClClCl

CAS RN 2745-49-5

IUPAC Name: 1,4-dichloro-2-(chloromethyl)benzene
Synonyms: 1,4-dichloro-2-(chloromethyl)benzene
Molecular Weight (g/mol): 195.47
Molecular Formula: C7H5Cl3
InChi Key: OMZINLIPPVNUOG-UHFFFAOYSA-N
SMILES: ClCC1=CC(Cl)=CC=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results