missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dichlorobenzyl chloride, 97%

Product Code. 11321937
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11321937

Brand: Thermo Scientific Alfa Aesar L07506.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2745-49-5 | C7H5Cl3 | 195.467 g/mol

2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichlorobenzyl chloride is used as pharmaceutical Intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2745-49-5
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
MDL Number MFCD00035818
InChI Key OMZINLIPPVNUOG-UHFFFAOYSA-N
Synonym 2,5-dichlorobenzyl chloride, 1,4-dichloro-2-chloromethyl benzene, 2,5-dichlorobenzylchloride, 1,4-dichloro-2-chloromethyl-benzene, unii-g46i9555fb, benzene, 1,4-dichloro-2-chloromethyl, acmc-20a58t, alpha,2,5-trichlorotoluene, unii-6q4qcu6n2r component
PubChem CID 75976
IUPAC Name 1,4-dichloro-2-(chloromethyl)benzene
SMILES C1=CC(=C(C=C1Cl)CCl)Cl

Specifications

Boiling Point 121°C to 124°C (14 mmHg)
Odor Pungent
Refractive Index 1.5755
Quantity 25 g
UN Number UN3265
Beilstein 637802
Solubility Information Not miscible in water.
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 2,5-Dichlorobenzyl chloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.