accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOHNO

CAS RN 2763-96-4

IUPAC Name: 5-(aminomethyl)-2,3-dihydro-1,2-oxazol-3-one
Synonyms: muscimol
Molecular Weight (g/mol): 114.1
Molecular Formula: C4H6N2O2
InChi Key: ZJQHPWUVQPJPQT-UHFFFAOYSA-N
SMILES: NCC1=CC(=O)NO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results