accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 27631-29-4

IUPAC Name: 2,4-dichloro-6,7-dimethoxyquinazoline
Synonyms: 2,4-dichloro-6,7-dimethoxyquinazoline
Molecular Weight (g/mol): 259.09
Molecular Formula: C10H8Cl2N2O2
InChi Key: DGHKCBSVAZXEPP-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(Cl)=NC(Cl)=NC2=C1
Molecular Weight (g/mol) 
  • (3)
  • (7)
Quantity 
  • (2)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results