CAS RN 2765-97-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Propinox Hydrochloride, TRC
CAS: 2765-97-1 Molecular Formula: C21 H23 N O3 . Cl H Molecular Weight (g/mol): 373.87 Synonym: Diphenyl(2-propynyloxy)acetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride,α-Phenyl-α-(2-propynyloxy)benzeneacetic Acid 2-(Dimethylamino)ethyl Ester Hydrochloride),Sertal,β-(Dimethylamino)ethyl α,α-diphenyl-α-(propargyloxy)acetate Hydrochloride,Pargeverine Hydrochloride IUPAC Name: 2-(dimethylamino)ethyl 2,2-diphenyl-2-prop-2-ynoxyacetate;hydrochloride SMILES: Cl.CN(C)CCOC(=O)C(OCC#C)(c1ccccc1)c2ccccc2