CAS RN 27893-41-0
IUPAC Name:
4-(heptyloxy)benzaldehyde
Synonyms:
4-(heptyloxy)benzaldehyde
Molecular Weight (g/mol):
220.31
Molecular Formula:
C14H20O2
InChi Key:
YBCKMIZXHKVONZ-UHFFFAOYSA-N
SMILES:
CCCCCCCOC1=CC=C(C=O)C=C1
1
–
1
of
1
results
1
4-n-Heptyloxybenzaldehyde, 97%
CAS: 27893-41-0 Molecular Formula: C14H20O2 Molecular Weight (g/mol): 220.312 MDL Number: MFCD00016616 InChI Key: YBCKMIZXHKVONZ-UHFFFAOYSA-N Synonym: 4-heptyloxybenzaldehyde,4-heptyloxy benzaldehyde,p-heptyloxybenzaldehyde,4-n-heptyloxybenzaldehyde,benzaldehyde, 4-heptyloxy,p-heptyloxy benzaldehyde,4-n-heptoxybenzaldehyde,4-heptyloxy-benzaldehyde,p-n-heptoxybenzaldehyde,acmc-209gzj PubChem CID: 119740 IUPAC Name: 4-heptoxybenzaldehyde SMILES: CCCCCCCOC1=CC=C(C=C1)C=O