CAS RN 27911-63-3
IUPAC Name:
(1R)-1-(pyridin-2-yl)ethan-1-ol
Synonyms:
(1R)-1-(pyridin-2-yl)ethanol
Molecular Weight (g/mol):
123.16
Molecular Formula:
C7H9NO
InChi Key:
PPHIIIRFJKDTLG-ZCFIWIBFSA-N
SMILES:
C[C@@H](O)C1=CC=CC=N1
1
–
2
of
2
results
2
(R)-2-(1-Hydroxyethyl)pyridine, 98%, Thermo Scientific Chemicals
CAS: 27911-63-3 Molecular Formula: C7H9NO Molecular Weight (g/mol): 123.15 MDL Number: MFCD04972322 InChI Key: PPHIIIRFJKDTLG-ZCFIWIBFSA-N Synonym: r-1-pyridin-2-yl ethanol,r-2-1-hydroxyethyl pyridine,r-1-2-pyridyl ethanol,1r-1-pyridin-2-yl ethan-1-ol,r-alpha-methyl-2-pyridinemethanol,1r-1-pyridin-2-ylethanol,1r-1-pyridin-2-yl ethanol,pubchem5699,r-1-pyridin-2-yl-ethanol,1r-1-2-pyridyl ethanol PubChem CID: 642847 IUPAC Name: (1R)-1-pyridin-2-ylethanol SMILES: CC(C1=CC=CC=N1)O