accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FHNO

CAS RN 2795-41-7

IUPAC Name: 6-fluoro-1H-indole-3-carbaldehyde
Synonyms: 6-fluoro-1H-indole-3-carbaldehyde
Molecular Weight (g/mol): 163.15
Molecular Formula: C9H6FNO
InChi Key: CWCYUOSLRVAKQZ-UHFFFAOYSA-N
SMILES: FC1=CC=C2C(NC=C2C=O)=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results