accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CCH3O(E)

CAS RN 2802-08-6

IUPAC Name: (3E)-4-(dimethylamino)but-3-en-2-one
Synonyms: (3E)-4-(dimethylamino)but-3-en-2-one
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: QPWSKIGAQZAJKS-SNAWJCMRSA-N
SMILES: CN(C)\C=C\C(C)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results