CAS RN 284028-89-3
CAS RN 284028-89-3
IUPAC Name:
2-[4-(trifluoromethyl)phenyl]-1H,4H,5H,7H,8H-thiopyrano[4,3-d]pyrimidin-4-one
Synonyms:
2-[4-(trifluoromethyl)phenyl]-1H,5H,7H,8H-thiopyrano[4,3-d]pyrimidin-4-one
Molecular Weight (g/mol):
312.31
Molecular Formula:
C14H11F3N2OS
InChi Key:
KLGQSVMIPOVQAX-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(C=C1)C1=NC(=O)C2=C(CCSC2)N1
1
–
5
of
5
results
1
Tocris Bioscience™ XAV 939
CAS: 284028-89-3 Molecular Formula: C14H11F3N2OS
2
XAV 939, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
XAV 939, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
XAV 939, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5