accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28479-22-3

CH3ClNCO

CAS RN 28479-22-3

IUPAC Name: 2-chloro-4-isocyanato-1-methylbenzene
Synonyms: 2-chloro-4-isocyanato-1-methylbenzene
Molecular Weight (g/mol): 167.59
Molecular Formula: C8H6ClNO
InChi Key: UKTKKMZDESVUEE-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1Cl)N=C=O
Boiling Point 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)