missing translation for 'onlineSavingsMsg'
Learn More

3-chloro-4-methylphenyl isocyanate, 98%, Thermo Scientific™

Product Code. 11985681 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11985681 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11985681 Supplier Thermo Scientific Acros Supplier No. 390430050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 23.0°C
CAS 28479-22-3
CAS Min % 97.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.2300g/mL
Boiling Point 225.0°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid or Low Melting Solid
Refractive Index 1.5560 to 1.5580 (20°C, 589nm)
Assay Percent Range 97.5% min. (GC)
Specific Gravity 1.23
Molecular Formula C8H6ClNO
MDL Number MFCD00013858
Synonym 3-chloro-4-methylphenyl isocyanate, 3-chloro-4-methylphenylisocyanate, benzene, 2-chloro-4-isocyanato-1-methyl, 3-chloro-p-tolyl isocyanate, isocyanic acid 3-chloro-4-methylphenyl ester, 3-chloro-4-methyl phenyl isocyanate, acmc-1ar7g, 3-chlor-4-methylphenylisocyanat, 2-chloro-4-isocyanato toluene, 3-chloro-4-methylphenyl isocyanate un2236 poison
Solubility Information Solubility in water: decomposes
InChI Key UKTKKMZDESVUEE-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1Cl)N=C=O
IUPAC Name 2-chloro-4-isocyanato-1-methylbenzene
Molecular Weight (g/mol) 167.59
PubChem CID 62832
Formula Weight 167.59
Percent Purity 98%
Chemical Name or Material 3-Chloro-4-methylphenyl isocyanate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Chloro-4-methylphenyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.