accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 28583-62-2

H3CONHCH3OHO

CAS RN 28583-62-2

IUPAC Name: N-(4-butanoyl-3-hydroxyphenyl)acetamide
Synonyms: N-(4-butanoyl-3-hydroxyphenyl)acetamide
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: RSBIKRQPSYDRFZ-UHFFFAOYSA-N
SMILES: CCCC(=O)C1=CC=C(NC(C)=O)C=C1O
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results