accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COBrOCH3

CAS RN 2859-78-1

IUPAC Name: 4-bromo-1,2-dimethoxybenzene
Synonyms: 4-bromo-1,2-dimethoxybenzene
Molecular Weight (g/mol): 217.06
Molecular Formula: C8H9BrO2
InChi Key: KBTMGSMZIKLAHN-UHFFFAOYSA-N
SMILES: COC1=CC=C(Br)C=C1OC
Molecular Weight (g/mol) 
  • (6)
  • (3)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (6)
  • (3)