accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CHNOOHO

CAS RN 2901-80-6

IUPAC Name: 3-methyl-2-(phenylformamido)butanoic acid
Synonyms: 3-methyl-2-(phenylformamido)butanoic acid
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: MIYQNOPLWKCHED-UHFFFAOYNA-N
SMILES: CC(C)C(NC(=O)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)