accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNH3CH3CFPFFFFF

CAS RN 292140-86-4

IUPAC Name: 1-ethyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate
Molecular Weight (g/mol): 270.16
Molecular Formula: C7H13F6N2P
InChi Key: XLKYEJKYOZNSDQ-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCN1C=C[N+](C)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results