missing translation for 'onlineSavingsMsg'
Learn More

1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate, 98%, Thermo Scientific™

Product Code. 15475576
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15475576 50 g 50g
15465576 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475576 Supplier Thermo Scientific Alfa Aesar Supplier No. H60228.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 292140-86-4
Molecular Formula C7H13F6N2P
Molecular Weight (g/mol) 270.159
MDL Number MFCD08458503
InChI Key XLKYEJKYOZNSDQ-UHFFFAOYSA-N
Synonym 1-ethyl-2,3-dimethylimidazolium hexafluorophosphate, 1-ethyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 3-ethyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, dsstox_cid_29120, dsstox_rid_83339, dsstox_gsid_49264, 1-ethyl-2,3-dimethylimidazol-1-ium hexafluorophosphate
PubChem CID 50878556
IUPAC Name 1-ethyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES CCN1C=C[N+](=C1C)C.F[P-](F)(F)(F)(F)F

Specifications

Quantity 50 g
Formula Weight 270.16
Percent Purity 98%
Chemical Name or Material 1-Ethyl-2,3-dimethylimidazolium hexafluorophosphate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.