CAS RN 29277-73-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile; (Atenolol Impurity H), TRC
CAS: 29277-73-4 Molecular Formula: C14 H20 N2 O2 Molecular Weight (g/mol): 248.32 Synonym: 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile,Atenolol Imp. H (EP) IUPAC Name: 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetonitrile SMILES: CC(C)NCC(O)COc1ccc(CC#N)cc1
2
Atenolol EP Impurity H (Atenolol USP Related Compound B), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.