missing translation for 'onlineSavingsMsg'
Learn More

2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile; (Atenolol Impurity H), TRC

Product Code. 30406929
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406929 100 mg -
30390844 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30406929 Supplier Toronto Research Chemicals Supplier No. TRCH946505100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29277-73-4
Molecular Formula C14 H20 N2 O2
Molecular Weight (g/mol) 248.32
Synonym 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile, Atenolol Imp. H (EP)
IUPAC Name 2-[4-[2-hydroxy-3-(propan-2-ylamino)propoxy]phenyl]acetonitrile
SMILES CC(C)NCC(O)COc1ccc(CC#N)cc1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H20N2O2/c1-11(2)16-9-13(17)10-18-14-5-3-12(4-6-14)7-8-15/h3-6,11,13,16-17H,7,9-10H2,1-2H3
Formula Weight 248.1525
Chemical Name or Material 2-[4-[(2RS)-2-Hydroxy-3-[(1-methylethyl)amino]propoxy]phenyl]acetonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.