accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOO(E/Z)

CAS RN 2960-66-9

IUPAC Name: ethyl 4-oxobut-2-enoate
Synonyms: ethyl 4-oxobut-2-enoate
Molecular Weight (g/mol): 128.13
Molecular Formula: C6H8O3
InChi Key: SDGAEBKMHIPSAC-UHFFFAOYSA-N
SMILES: CCOC(=O)C=CC=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results