accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOBrCl

CAS RN 29682-39-1

IUPAC Name: 1-bromo-2-chloro-4-nitrobenzene
Synonyms: 1-bromo-2-chloro-4-nitrobenzene
Molecular Weight (g/mol): 236.45
Molecular Formula: C6H3BrClNO2
InChi Key: YKSXEJZFIQAUHJ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Br)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results