missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-2-chloro-4-nitrobenzene, 98%

Product Code. 11341957
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341957 50 g 50g
11331957 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341957 Supplier Thermo Scientific Alfa Aesar Supplier No. L07573.18
 Restricted Item
Estimated Shipment: 24-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 29682-39-1 | C6H3BrClNO2 | 236.449 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29682-39-1
Molecular Formula C6H3BrClNO2
Molecular Weight (g/mol) 236.449
MDL Number MFCD00051514
InChI Key YKSXEJZFIQAUHJ-UHFFFAOYSA-N
Synonym 4-bromo-3-chloronitrobenzene, benzene,1-bromo-2-chloro-4-nitro, pubchem22601, acmc-209vlc, 2-chloro-4-nitro-bromobenzene, 4-bromo-3-chloro-nitrobenzene, bromo-2-chloro-4-nitrobenzene, 1-bromanyl-2-chloranyl-4-nitro-benzene
PubChem CID 34667
IUPAC Name 1-bromo-2-chloro-4-nitrobenzene
SMILES C1=CC(=C(C=C1[N+](=O)[O-])Cl)Br

Specifications

Melting Point 59°C to 62°C
Quantity 50 g
UN Number UN2811
Beilstein 2253107
Formula Weight 236.45
Percent Purity 98%
Chemical Name or Material 1-Bromo-2-chloro-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.