CAS RN 29702-43-0
IUPAC Name:
2-fluoro-3,4,5,6-tetrahydroxyhexanal
Synonyms:
2-fluoro-3,4,5,6-tetrahydroxyhexanal
Molecular Weight (g/mol):
182.15
Molecular Formula:
C6H11FO5
InChi Key:
AOYNUTHNTBLRMT-UHFFFAOYNA-N
SMILES:
OCC(O)C(O)C(O)C(F)C=O
1
–
1
of
1
results
1
2-Deoxy-2-fluoro-D-glucose, TRC
CAS: 29702-43-0 Molecular Formula: C6 H11 F O5 Molecular Weight (g/mol): 182.15 Synonym: 2-Deoxy-2-fluoro-D-glucose,2-Deoxy-2-fluoro-D-glucopyranose,2-Deoxy-2-fluoroglucose,2-Fluoro-2-deoxy-D-glucose,FDG,Fludeoxyglucose IUPAC Name: (2R,3S,4R,5R)-2-fluoro-3,4,5,6-tetrahydroxyhexanal SMILES: OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](F)C=O