CAS RN 29858-07-9
IUPAC Name:
magnesium(2+) ethoxyethane dibromide
Synonyms:
magnesium(2+) diethyl ether dibromide
Molecular Weight (g/mol):
258.24
Molecular Formula:
C4H10Br2MgO
InChi Key:
JGZKUKYUQJUUNE-UHFFFAOYSA-L
SMILES:
[Mg++].[Br-].[Br-].CCOCC
1
–
2
of
2
results
1
Promotions Available
Magnesium bromide ethyl etherate, 99%
Magnesium bromide ethyl etherate, 99%
CAS: 29858-07-9 Molecular Formula: C4H10Br2MgO Molecular Weight (g/mol): 258.24 MDL Number: MFCD00064500 InChI Key: JGZKUKYUQJUUNE-UHFFFAOYSA-L Synonym: magnesium bromide diethyl etherate,magnesium bromide etherate,magnesium bromide ethyl etherate,grignard reagent PubChem CID: 11010650 IUPAC Name: magnesium;ethoxyethane;dibromide SMILES: Br[Mg]Br.CCOCC