accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3010-80-8

HOClClCl

CAS RN 3010-80-8

IUPAC Name: tris(4-chlorophenyl)methanol
Synonyms: tris(4-chlorophenyl)methanol
Molecular Weight (g/mol): 363.66
Molecular Formula: C19H13Cl3O
InChi Key: BPFKTJMHOWDJKI-UHFFFAOYSA-N
SMILES: OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results