missing translation for 'onlineSavingsMsg'
Learn More

4,4',4″-Trichlorotrityl alcohol, 95%

Product Code. 11347076
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347076 10 g 10g
11337076 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11347076 Supplier Thermo Scientific Alfa Aesar Supplier No. L06965.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

CAS: 3010-80-8 | C19H13Cl3O | 363.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3010-80-8
Molecular Formula C19H13Cl3O
Molecular Weight (g/mol) 363.66
MDL Number MFCD00051795
InChI Key BPFKTJMHOWDJKI-UHFFFAOYSA-N
Synonym tris 4-chlorophenyl methanol, 4,4',4-trichlorotrityl alcohol, tris-4-chloro-phenyl-methanol, tri 4-chlorophenyl methanol, acmc-1clfy, tris-p-chlorophenyl methanol, tris 4-chlorophenyl methan-1-ol, 4,4,4-trichlorotrityl alcohol, benzenemethanol,4-chloro-a,a-bis 4-chlorophenyl
PubChem CID 76379
IUPAC Name tris(4-chlorophenyl)methanol
SMILES OC(C1=CC=C(Cl)C=C1)(C1=CC=C(Cl)C=C1)C1=CC=C(Cl)C=C1

Specifications

Melting Point 94°C to 97°C
Quantity 10 g
Beilstein 2293156
Formula Weight 363.67
Percent Purity 95%
Chemical Name or Material 4,4',4''-Trichlorotrityl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.