accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3047-38-9

IUPAC Name: cyclopent-1-ene-1-carbonitrile
Synonyms: cyclopent-1-ene-1-carbonitrile
Molecular Weight (g/mol): 93.13
Molecular Formula: C6H7N
InChi Key: XBVZRFXCDCYXAX-UHFFFAOYSA-N
SMILES: N#CC1=CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results