missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclopentenecarbonitrile, 95%

Product Code. 11475560
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11475560 5 g 5g
11465560 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475560 Supplier Thermo Scientific Alfa Aesar Supplier No. H25836.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 3047-38-9 | C6H7N | 93.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3047-38-9
Molecular Formula C6H7N
Molecular Weight (g/mol) 93.129
MDL Number MFCD00043765
InChI Key XBVZRFXCDCYXAX-UHFFFAOYSA-N
Synonym 1-cyanocyclopentene, cyclopent-1-enecarbonitrile, 1-cyclopentenecarbonitrile, 1-cyclopentene-1-carbonitrile, cyclopent-1-ene-1-carbonitrile, 1-cyanocyclopenten, acmc-20aovl, 1-cyclopentenyl cyanide, 1-cyano-1-cyclopentene
PubChem CID 137808
IUPAC Name cyclopentene-1-carbonitrile
SMILES C1CC=C(C1)C#N

Specifications

Density 0.944
Boiling Point 72°C (15 mmHg)
Refractive Index 1.4715
Quantity 5 g
UN Number UN3276
Formula Weight 93.14
Percent Purity 97%
Chemical Name or Material 1-Cyclopentenecarbonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.