accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 30595-79-0

OHClCl

CAS RN 30595-79-0

IUPAC Name: 2-(2,6-dichlorophenyl)ethan-1-ol
Synonyms: 2-(2,6-dichlorophenyl)ethanol
Molecular Weight (g/mol): 191.05
Molecular Formula: C8H8Cl2O
InChi Key: ZBQPKQUIKJDGIX-UHFFFAOYSA-N
SMILES: OCCC1=C(Cl)C=CC=C1Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)