accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2HOO

CAS RN 3060-50-2

IUPAC Name: 2-amino-2,2-diphenylacetic acid
Synonyms: aminodiphenylacetic acid
Molecular Weight (g/mol): 227.26
Molecular Formula: C14H13NO2
InChi Key: YBONNYNNFBAKLI-UHFFFAOYSA-N
SMILES: NC(C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results