accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31224-43-8

NOF

CAS RN 31224-43-8

IUPAC Name: 3-fluoropyridine-2-carbaldehyde
Synonyms: 3-fluoropyridine-2-carbaldehyde
Molecular Weight (g/mol): 125.1
Molecular Formula: C6H4FNO
InChi Key: OZIMPUNGBUYCSP-UHFFFAOYSA-N
SMILES: FC1=C(C=O)N=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (5)
  • (3)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (5)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)