accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 312700-07-5

NNH2HCl

CAS RN 312700-07-5

IUPAC Name: 4-(9H-carbazol-9-yl)aniline hydrochloride
Synonyms: 4-(carbazol-9-yl)aniline hydrochloride
Molecular Weight (g/mol): 294.78
Molecular Formula: C18H15ClN2
InChi Key: CFQROJKMLJRSAL-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=C(C=C1)N1C2=C(C=CC=C2)C2=C1C=CC=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results