missing translation for 'onlineSavingsMsg'
Learn More

9-(4-Aminophenyl)carbazole hydrochloride, 98%

Product Code. 11367736
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11367736 250 mg 250mg
11357736 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11367736 Supplier Thermo Scientific Alfa Aesar Supplier No. L11362.MD
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 312700-07-5 | C18H15ClN2 | 294.782 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 312700-07-5
Molecular Formula C18H15ClN2
Molecular Weight (g/mol) 294.782
MDL Number MFCD00082678
InChI Key CFQROJKMLJRSAL-UHFFFAOYSA-N
Synonym 4-9h-carbazol-9-yl aniline hydrochloride, 9-4-aminophenyl carbazole hydrochloride, 4-carbazol-9-yl aniline hydrochloride, 4-carbazol-9-ylaniline hydrochloride, acmc-1agoy
PubChem CID 2794996
IUPAC Name 4-carbazol-9-ylaniline;hydrochloride
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3N2C4=CC=C(C=C4)N.Cl

Specifications

Melting Point ∽250°C (decomposition)
Quantity 250 mg
Formula Weight 294.79
Percent Purity 98%
Chemical Name or Material 9-(4-Aminophenyl)carbazole hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.