CAS RN 31317-07-4
CAS RN 31317-07-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Methyldibenzothiophene, TRC
CAS: 31317-07-4 Molecular Formula: C13 H10 S Molecular Weight (g/mol): 198.28 Synonym: 1-Methyldibenzo[b,d]thiophene; IUPAC Name: 1-methyldibenzothiophene SMILES: Cc1cccc2sc3ccccc3c12