missing translation for 'onlineSavingsMsg'
Learn More

1-Methyldibenzothiophene, TRC

Product Code. 30133832
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133832 100 mg 100mg
30009095 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133832 Supplier Toronto Research Chemicals Supplier No. TRCM299750100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31317-07-4
Molecular Formula C13 H10 S
Molecular Weight (g/mol) 198.28
Synonym 1-Methyldibenzo[b,d]thiophene;
IUPAC Name 1-methyldibenzothiophene
SMILES Cc1cccc2sc3ccccc3c12

Specifications

InChI Formula InChI=1S/C13H10S/c1-9-5-4-8-12-13(9)10-6-2-3-7-11(10)14-12/h2-8H,1H3
Quantity 100 mg
Formula Weight 198.0503
Shipping Condition Room Temperature
Chemical Name or Material 1-Methyldibenzothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.