accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 31333-13-8

H3COH

CAS RN 31333-13-8

IUPAC Name: non-3-yn-1-ol
Synonyms: non-3-yn-1-ol
Molecular Weight (g/mol): 140.23
Molecular Formula: C9H16O
InChi Key: TZZVRLFUTNYDEG-UHFFFAOYSA-N
SMILES: CCCCCC#CCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)