missing translation for 'onlineSavingsMsg'
Learn More

3-Nonyn-1-ol, 98%, Thermo Scientific™

Product Code. 11459743
Click to view available options
Quantity:
5 g
10 g
Unit Size:
10g
5g
This item is not returnable. View return policy

Product Code. 11459743

Brand: Thermo Scientific Alfa Aesar A12922.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in the synthesis of 3,3,4,4- 2 H 4 -nonanol and 3,3,4,4- 2 H 4 -nonanal.

Solubility
Optimally soluble in aqueous solutions.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31333-13-8
Molecular Formula C9H16O
Molecular Weight (g/mol) 140.23
MDL Number MFCD00002958
InChI Key TZZVRLFUTNYDEG-UHFFFAOYSA-N
Synonym 3-nonyn-1-ol, 2-hydroxyethyl pentylacetylene, 1-hydroxy-3-nonin, pubchem13073, acmc-209hlo
PubChem CID 123437
IUPAC Name non-3-yn-1-ol
SMILES CCCCCC#CCCO

Specifications

Density 0.884
Boiling Point 111°C to 112°C (20 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.457
Quantity 5 g
Beilstein 1700444
Solubility Information Optimally soluble in aqueous solutions.
Formula Weight 140.23
Percent Purity 98%
Chemical Name or Material 3-Nonyn-1-ol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.