accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3153-26-2

IUPAC Name: oxovanadium; bis(pentane-2,4-dione)
Synonyms: vanadyl acetylacetonate
Molecular Weight (g/mol): 267.18
Molecular Formula: C10H16O5V
InChi Key: GGUKAPURIDGXAJ-UHFFFAOYSA-N
SMILES: O=[V].CC(=O)CC(C)=O.CC(=O)CC(C)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results