accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3158-26-7

IUPAC Name: 1-isocyanatooctane
Synonyms: 1-isocyanatooctane
Molecular Weight (g/mol): 155.24
Molecular Formula: C9H17NO
InChi Key: DYQFCTCUULUMTQ-UHFFFAOYSA-N
SMILES: CCCCCCCCN=C=O
Molecular Weight (g/mol) 
  • (6)
  • (2)
Percent Purity 
  • (6)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (6)
  • (2)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)
  • (4)
  • (2)