missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isocyanate, 97%, Thermo Scientific Chemicals

Product Code. 11309515
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11309515 5 g 5g
11399505 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309515 Supplier Thermo Scientific Alfa Aesar Supplier No. L18746.06
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3158-26-7 | Molecular Formula: C9H17NO | Molecular Weight: 155.241 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3158-26-7
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.241
MDL Number MFCD00013887
InChI Key DYQFCTCUULUMTQ-UHFFFAOYSA-N
Synonym octyl isocyanate, octylisocyanate, octane, 1-isocyanato, n-octylisocyanate, 1-octyl isocyanate, n-octyl isocyanate, 1-isocyanato-octane, isocyanic acid, octyl ester, octanisocyanate
PubChem CID 76616
IUPAC Name 1-isocyanatooctane
SMILES CCCCCCCCN=C=O

Specifications

Density 0.88
Boiling Point 200°C to 204°C
Flash Point 71°C (159°F)
Odor Pungent
Refractive Index 1.432
Quantity 5 g
UN Number UN2206
Sensitivity Moisture sensitive
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-Octyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.