CAS RN 31599-60-7
IUPAC Name:
1-iodo-2,3-dimethylbenzene
Synonyms:
1-iodo-2,3-dimethylbenzene
Molecular Weight (g/mol):
232.06
Molecular Formula:
C8H9I
InChi Key:
DANMWBNOPFBJSZ-UHFFFAOYSA-N
SMILES:
CC1=CC=CC(I)=C1C
1
–
2
of
2
results
1
3-Iodo-o-xylene, 97%
CAS: 31599-60-7 Molecular Formula: C8H9I Molecular Weight (g/mol): 232.064 MDL Number: MFCD00041437 InChI Key: DANMWBNOPFBJSZ-UHFFFAOYSA-N Synonym: 3-iodo-o-xylene,1,2-dimethyl-3-iodobenzene,2,3-dimethyliodobenzene,benzene, 1-iodo-2,3-dimethyl,benzene, iododimethyl,2,3-dimethyl-1-iodobenzene,jodxylol,3-iodo-ortho-xylene,pubchem3865,acmc-1ajf2 PubChem CID: 141645 IUPAC Name: 1-iodo-2,3-dimethylbenzene SMILES: CC1=C(C(=CC=C1)I)C