missing translation for 'onlineSavingsMsg'
Learn More

3-Iodo-o-xylene, 98%

Product Code. 10711351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
Packaging:
Glass bottle
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711351 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10711351 Supplier Thermo Scientific Acros Supplier No. 309350050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 31599-60-7 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 31599-60-7
CAS Min % 97.5
Color Brown to Brown-Red
CAS Max % 100.0
Density 1.64g/mL
Quantity 5 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.6060 to 1.6080 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C8H9I
Linear Formula (CH3)C6H3I
MDL Number MFCD00041437
Synonym 3-iodo-o-xylene, 1,2-dimethyl-3-iodobenzene, 2,3-dimethyliodobenzene, benzene, 1-iodo-2,3-dimethyl, benzene, iododimethyl, 2,3-dimethyl-1-iodobenzene, jodxylol, 3-iodo-ortho-xylene, pubchem3865, acmc-1ajf2
InChI Key DANMWBNOPFBJSZ-UHFFFAOYSA-N
SMILES CC1=C(C(=CC=C1)I)C
IUPAC Name 1-iodo-2,3-dimethylbenzene
Molecular Weight (g/mol) 232.064
PubChem CID 141645
Formula Weight 232.07
Percent Purity ≥97.5%
Chemical Name or Material 3-Iodo-o-xylene, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.