accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH3CNHH3CH3CCH3OCl

CAS RN 31677-93-7

IUPAC Name: hydrogen 2-(tert-butylamino)-1-(3-chlorophenyl)propan-1-one chloride
Synonyms: hydrogen bupropion chloride
Molecular Weight (g/mol): 276.2
Molecular Formula: C13H19Cl2NO
InChi Key: HEYVINCGKDONRU-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CC(NC(C)(C)C)C(=O)C1=CC=CC(Cl)=C1
Molecular Weight (g/mol) 
  • (7)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Industry Type 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (7)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)