accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3189-22-8

H3COHN

CAS RN 3189-22-8

IUPAC Name: 7-methoxy-1H-indole
Synonyms: 7-methoxy-1H-indole
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: FSOPPXYMWZOKRM-UHFFFAOYSA-N
SMILES: COC1=C2NC=CC2=CC=C1
Boiling Point 
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Physical Form 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)