accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 3202-33-3

NHO

CAS RN 3202-33-3

IUPAC Name: 4-phenoxypiperidine
Synonyms: 4-phenoxypiperidine
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: KBYPITRKIJKGMD-UHFFFAOYSA-N
SMILES: C1CC(CCN1)OC1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)