accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 32022-38-1

CH3ClOONHNH2

CAS RN 32022-38-1

IUPAC Name: 2-(4-chloro-2-methylphenoxy)acetohydrazide
Synonyms: 2-(4-chloro-2-methylphenoxy)acetohydrazide
Molecular Weight (g/mol): 214.65
Molecular Formula: C9H11ClN2O2
InChi Key: JXQVZCTYFYWQGN-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC=C1OCC(=O)NN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results