missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide, 96%, Thermo Scientific™

Product Code. 11450348
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11450348 25 g 25g
11440348 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450348 Supplier Thermo Scientific Alfa Aesar Supplier No. B24521.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32022-38-1
Molecular Formula C9H11ClN2O2
Molecular Weight (g/mol) 214.65
MDL Number MFCD00082720
InChI Key JXQVZCTYFYWQGN-UHFFFAOYSA-N
Synonym 2-4-chloro-2-methylphenoxy acetohydrazide, 2-4-chloro-2-methylphenoxy acetic acid hydrazide, acetic acid, 4-chloro-o-tolyl oxy-, hydrazide, acmc-20aopt, 4-chloro-o-tolyl oxy acetic acid hydrazide, 2-4-chloro-2-methyl-phenoxy acetohydrazide, 2-4-chloro-3-methylphenoxy acetic hydrazide, 2-4-chloro-o-tolyloxy acetic acid hydrazide, 2-methyl-4-chlorophenoxyacetic acid hydrazide, 4-chloro-2-methylphenoxy acetic acid hydrazid
PubChem CID 149463
IUPAC Name 2-(4-chloro-2-methylphenoxy)acetohydrazide
SMILES CC1=CC(Cl)=CC=C1OCC(=O)NN

Specifications

Melting Point 149°C to 151°C
Quantity 25 g
Beilstein 983137
Formula Weight 214.65
Percent Purity 96%
Chemical Name or Material 2-(4-Chloro-2-methylphenoxy)acetic acid hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.